Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

epithelial Sodium Channel gamma Mouse anti-Human, Alexa Fluor™ 532, Clone: 11-35-1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP241374AF532

Catalog No. NP241374A32


Add to cart

Description

Description

epithelial Sodium Channel gamma Monoclonal antibody specifically detects epithelial Sodium Channel gamma in Human samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications

Specifications

epithelial Sodium Channel gamma
11-35-1
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
amiloride-sensitive epithelial sodium channel gamma subunit, amiloride-sensitive sodium channel gamma-subunit, BESC3, ENaC gamma subunit, ENaCG, ENaCgamma, Epithelial Na(+) channel subunit gamma, gamma-ENaC, gamma-NaCH, Nonvoltage-gated sodium channel 1 subunit gamma, PHA1, SCNEGamiloride-sensitive sodium channel subunit gamma, sodium channel, nonvoltage-gated 1, gamma
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Antibody is specific for the neoepitope generated after proteolytic cleavage of human gENaC and recognizes the C-terminal fragment starting from amino acid residue 182.
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
SCNN1G
Synthetic peptide seguence: VGGSIIHKAC
0.1 mL
Primary
6340
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit