Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence, flow cytometry and radioimaging experiments.

Specifications

Antigen Epithelial Specific Antigen/Ep-CAM Ab-3
Clone 323/A3
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS181P
View Documents
Thermo Scientific™
MS181P
Purified Ab with BSA and Azide 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS181P1
View Documents
Thermo Scientific™
MS181P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence, flow cytometry and radioimaging experiments with Thermo Scientific™ Epithelial Specific Antigen/Ep-CAM Ab-3, Mouse Monoclonal Antibody.

EGP40 is a 40 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. It reportedly distinguishes adenocarcinomas from pleural mesotheliomas.

Host Species: Mouse

Clone: 323/A3

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: MCF-7 human breast cancer cells

Molecular Weight: 40-43 kDa

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry
  • Radioimaging

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Epithelial Specific Antigen/Ep-CAM Ab-3
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Murine
Cancer and Tumor Biology
MCF-7 human breast cancer cells
323/A3
Monoclonal
Concentrated
RUO
Primary
IgG1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit