Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS1825R7

 View more versions of this product

Catalog No. MS1825R7


Add to cart

Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: MOC-31

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1)

Molecular Weight: 40 kDa

Positive Control: Colon CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Epithelial Specific Antigen/Ep-CAM Ab-7
Monoclonal
Unconjugated
Murine
IgG2a kappa
RUO
Primary
Monoclonal
Immunohistochemistry (Paraffin)
MOC-31
Prediluted
Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1)
7mL
Cancer and Tumor Biology
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit