Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody.

$135.00 - $644.00

Specifications

Antigen Epithelial Specific Antigen/Ep-CAM Ab-7
Clone MOC-31
Immunogen Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1)
Monoclonal or Polyclonal Monoclonal
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS1825R7
View Documents
Thermo Scientific™
MS1825R7
7mL
Each for $401.00
Add to cart
 
MS1825S
View Documents
Thermo Scientific™
MS1825S
1mL
Each for $644.00
Add to cart
 
MS1825S0
View Documents
Thermo Scientific™
MS1825S0
100μL
Each for $135.00
Add to cart
 
MS1825R1
View Documents
Thermo Scientific™
MS1825R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: MOC-31

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1)

Molecular Weight: 40 kDa

Positive Control: Colon CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Epithelial Specific Antigen/Ep-CAM Ab-7
Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1)
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
MOC-31
Monoclonal
Monoclonal
Murine
Cancer and Tumor Biology
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit