Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPLIN Mouse anti-Human, Clone: OTI9F3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527021

Catalog No. PIMA527021


Add to cart

Description

Description

EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles (Maul et al., 2003 [PubMed 12566430]).
Specifications

Specifications

EPLIN
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UHB6
EPLIN; SREBP3; 1110021C24Rik; LIM domain and actin-binding protein 1; epithelial protein lost in neoplasm beta; sterol regulatory element binding protein 3
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51474
Immunohistochemistry (Paraffin), Western Blot
OTI9F3
Unconjugated
LIMA1
Liquid
LIMA1
Human recombit protein fragment corresponding to amino acids 1-275 of LIMA1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit