Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPM2AIP1 Mouse anti-Human, Clone: OTI2G3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501797

Catalog No. 50-167-2913


Add to cart

Description

Description

The EPM2A gene, which encodes laforin, is mutated in an autosomal recessive form of adolescent progressive myoclonus epilepsy. The protein encoded by this gene binds to laforin, but its function is not known. This gene is intronless.
Specifications

Specifications

EPM2AIP1
Monoclonal
0.7 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q7L775
EPM2A interacting protein 1
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9852
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2G3
Unconjugated
EPM2AIP1
Liquid
EPM2AIP1
Full length human recombit protein of human EPM2AIP1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit