Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPM2AIP1 Mouse anti-Human, PE-Cyanine7, Clone: OTI2A2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271497PECY7

Catalog No. N271497PEC7


Add to cart

Description

Description

EPM2AIP1 Monoclonal antibody specifically detects EPM2AIP1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

EPM2AIP1
OTI2A2
Flow Cytometry
Affinity Purified, Conjugated
EPM2AIP1
Full length recombinant protein of human EPM2AIP1 (NP_055620) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
EPM2A (laforin) interacting protein 1, EPM2A interacting protein 1, EPM2A-interacting protein 1, KIAA0766FLJ11207, Laforin-interacting protein
Mouse
IgG2b
0.1 mL
Primary
9852.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit