Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPM2AIP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549324

Catalog No. PA549324

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The EPM2A gene, which encodes laforin, is mutated in an autosomal recessive form of adolescent progressive myoclonus epilepsy. The protein encoded by this gene binds to laforin, but its function is not known. This gene is intronless. [provided by RefSeq, Oct 2008]
Specifications

Specifications

EPM2AIP1
Polyclonal
PBS with 0.09% sodium azide
Q7L775, Q8VEH5
EPM2A-interacting protein 1, Laforin-interacting protein, EPM2AIP1, KIAA0766
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
77781, 9852
Western Blot
Unconjugated
EPM2AIP1
Liquid
EPM2AIP1
KLH conjugated synthetic peptide between 39-62 amino acids from the N-terminal region of human EPM2AIP1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit