Learn More
Aviva Systems Biology Corp EPN2 Antibody - middle region (ARP55480_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55480_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EPN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EPN2. Alternate names: EHB21, KIAA1065ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61519