Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPO Mouse anti-Human, Clone: EPO/1368, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14518
Description
Mouse monoclonal antibody raised against partial recombinant human EPO.
- High specificity
- High reactivity
Specifications
EPO | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by | |
P01588 | |
EPO | |
Recombinant protein corresponding to amino acids 28-162 of human EPO. | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
EPO/1368 | |
Mouse monoclonal antibody raised against partial recombinant human EPO. | |
In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
EP/MGC138142 | |
Mouse | |
Protein A/G purification | |
RUO | |
2056 | |
Store at 4°C. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction