Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Epo Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA130182

Catalog No. PA130182


Only null left
Add to Cart

Description

Description

PA1-30182 has been tested for its ability to neutralize the biological activity of EPO on the human cell line, TF1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Epo
Polyclonal
Unconjugated
EPO
EP; EPO; Epoetin; erythropoietin; MGC138142; MVCD2
Rabbit
IgG fraction
RUO
2056
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Inhibition Assays, Immunohistochemistry (Paraffin), Neutralization, Western Blot
1 mg/mL
PBS with 5% trehalose and no preservative
P01588
EPO
Recombinant full length erythropoietin (Human), expressed in CHO cells.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.