Learn More
Selleck Chemical LLCSupplier Diversity Partner Epothilone A 5mg 152044-53-6  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S12975MG
Epothilone A is a paclitaxel-like microtubule-stabilizing agent with EC0.01 of 2 μM.
Catalog No. 50-712-0