Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epothilone B, Tocris Bioscience™
Click to view available options
Quantity:
100 μg
Description
Microtubule stabilization agent that promotes tubulin polymerization and induces G2-M cell cycle arrest (EC50 = 32 nM in HeLa cells). Inhibits proliferation of human carcinoma cell lines in vitro, including MDR cells overexpressing the P-glycoprotein efflux pump. Exhibits potent cytotoxicity in MCF-7 and A549 cells (EC50 values are 0.3 and 2.7 nM respectively). Inhibits growth of HCT-15 tumors in mice in vivo.
Specifications
Quantity | 100 μg |
Formula Weight | 507.68 |
Percent Purity | >98% |
Chemical Name or Material | Epothilone B |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction