Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ 30-Place Fixed-Angle Rotors
Click to view available options
Includes:
Aerosol-tight lid
Quick release lid
Description
Includes
Cover
Ideal for containing hazardous materials such as HIV, TB and radioactive samples.
- Accept up to 30 × 1.5 to 2.0mL tubes directly or 0.5mL, 0.25/0.4mL, Microtainer™ and 0.2mL PCR tubes with accessory adapters
- Maximum speed/RCF: 14,000rpm/20,800 x g
- Available with quick-release lid or aerosol-tight cover

Specifications
Specifications
Type | Fixed Angle Rotor |
Capacity | 30 x 1.5/2 mL |
Max. RCF | 20,800 x g |
Max. Speed | 14,000 rpm |
For Use With (Equipment) | Eppendorf 5417C/5417R Microcentrifuge |
Angle | Fixed |
Includes | Quick release lid |
Autoclavable | Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction