Eppendorf™ 5702 Series Centrifuge Available on GSA/VA Contract for Federal Government customers only.

Low speed, general purpose units designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs

Manufacturer: Eppendorf™ 022629916

 View more versions of this product

Catalog No. 05-413-240

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Max. Speed 4400rpm
    Noise Level < 45 dB(A) with Rotor A-4-38
    Wattage 380w
    Dimensions (L x W x H) Exterior 58 x 38 x 26cm (22.8 x 15 x 10.2 in.)
    Safety Features SOFT brake function
    Height (Metric) Open Cover 60cm (23.6 in.)
    Timer 0 – 99 min, with continuous run function
    Weight (English) 79.4 lb.
    Capacity (Metric) Max. Load 4 x 100mL
    Max. RCF 3000 x G
    Product Type Refrigerated
    Weight (Metric) 36kg
    Electrical Requirements 230V 50/60Hz

    Choose either the 5702 model - a base, non-heated centrifuge for standard applications, the 5702 R that is a refrigerated model for heat-sensitive samples, or the heated and refrigerated 5702 RH model for centrifugation under biological conditions to improve cell viability for more accurate results in downstream application like cell cultivation.
    Features

    • Max. speed: 3,000 x g (4,400 rpm)
    • Very compact footprint to fit on every lab bench
    • Very quiet operation to benefit your environment
    • Low access height for simple loading and unloading of samples
    • SOFT brake function for slow acceleration and braking. Optimized for cell separation by gradient centrifugation
    • “At set RPM” function starts the timer when the selected rotational speed is reached for reproducible centrifugation runs
    • Key lock to prevent unintentional adjustments
    • Stainless steel rotor chamber is rust-free and easy to clean
    • Electronic imbalance detection for maximum safety
    • Optimizes air flow minimizes sample warming during run (5702)
    • FastTemp function for fast precooling (5702 R/RH)
    • Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)
    • ECO shut–off engages after 8 hours of nonuse to reduce energy consumption and extend compressor life (5702 R/RH)
    • Stainless steel rotor chamber is rust-free and easy to clean
    • Imbalance detection for maximum safety
    • Key lock to prevent accidental parameter changes
    • Aerosol–tight caps for safe centrifugation of hazardous samples optionally available

    Additional Features for 5702 R and 5702 RH
    • Temperature settings from -9 to 40°C
    • FastTemp™ function for fast precooling
    • Standby cooling keeps the 5702 R at set temperature when lid is closed
    • ECO shut-off engages after 8 hours of non-use to reduce energy consumption and extend compressor life
    • Two program keys to store routine procedures
    • Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)

    Packages
    All packages include one centrifuge and a rotor. Other parts included may be a bucket, adapters, and rotary knobs. See ordering table for specifics of each package.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit