Eppendorf™ 5702 Series Fixed-Angle Rotors Available on GSA/VA Contract for Federal Government customers only.
For use with 5702 centrifuge
Manufacturer: Eppendorf™ 022639421
 View more versions of this product
Catalog No. 05-400-305
  Check Availability
Add to Cart
    Specifications
    Max. RCF 2750
    Speed 4400rpm
    Type Fixed angle rotor
    Capacity (Metric) 10 x 15mL
    For Use With (Equipment) 5702 Centrifuge
    Includes 10 Adapters and 10 Rubber mats, 10 Steel adapter sleeves, 10 cushions for conical tubes and 10 cushions for 15mL round bottom tubes
  • Description & Specifications
Related Products
Product Content Feedback