Learn More
Eppendorf™ 8-Place Swinging Bucket Rotor for Models 5417C/5417R Microcentrifuges
8 × 1.5 to 2.0mL capacity
Supplier:  Eppendorf™ 022636138
Catalog No. 05-406-41
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Includes: Rotor and cover
Description
Description
Provides flat pelletization for DNA precipitations and immunoprecipitations.
  • For use with 5417 series centrifuges
  • Max. speed/RCF: 10,000rpm/10,600 × g
Order Info
Using accessory adapters (sold separately), the rotor can accommodate 0.5mL, 0.25/0.4mL, Microtainer™ and 0.2mL PCR tubes.
Specifications
Specifications
Swinging Bucket Rotor
10,600 x g
Models 5417C/5417R
8 x 1.5/2 mL
10,000 rpm
Aluminum lid
SDS
Documents
Documents
Product Certifications
Accessories