Learn More
Eppendorf™ A-4-38 Rotor Adapters
For round and rectangular buckets
Supplier: Eppendorf™ 5702763004
Description
- For use with Centrifuge 5702/ 5702 R/5702 RH and A-4-38 Rotor
- Designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs.
Specifications
Rotor Adapter | |
9 x 5 mL Tubes | |
A-4-38 |