Learn More
Eppendorf™ Reference™ 2 Pipette Accessory, TrackIT Data Reader Available on GSA/VA Contract for Federal Government customers only.
Simple identification and documentation
Manufacturer:  Eppendorf™ 3903000014
Catalog No. 05-412-456
Add to cart
Description
Description
  • Product contains an RFID reader and software to scan the device data of an embedded RFID chip within liquid handling instruments
  • Saves data in a database which can be opened at any time
  • Device data can be exported individually or automatically in various formats
  • In combination with premium products like Eppendorf Reference 2, offers also the ability to add your own data on the chip like calibration time, location, special application or other important data
  • Exports information to several formats (HTML, XML and CVS) to implement the system into existing software like electronic lab book or LIMS
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
RFID Reader and Software
Instrument identification, traceability used in an experiment, individualization of the instrument
Reference 2 Pipets