Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Adapters for 6 x 15/50mL Tube Rotor
Rotor adapters offered in various sizes
Supplier: Eppendorf™ 022654567
Description
- Round
- Small
Specifications
Microcentrifuge Tube Adapter | |
29 mm | |
Eppendorf™ fixed angle rotor for microcentrifuge |
Round Bottom | |
95 to 125 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction