Eppendorf™ Biopur™ Racks and Singles Available on GSA/VA Contract for Federal Government customers only.
For the most stringent laboratory requirements
Manufacturer: Eppendorf™ 022491113
 View more versions of this product
Catalog No. 05-403-12
  Check Availability
Add to Cart
    Specifications
    Product Type Pipet Tips
    Material Polypropylene
    Length (English) 2.99 in.
    Length (Metric) 7.6cm
    Sterility Sterile
    Autoclavable Yes
    Tip Style Drop Contour Design, Extended, Finely Tapered, Universal Fit
    Packaging Racks, 5 x 96
    Certifications/Compliance ISO 9001
    Volume (Metric) 50 to 1250μL
    Color Green
  • Description & Specifications
Related Products
Product Content Feedback