Eppendorf™ Biopur™ Racks and Singles Available on GSA/VA Contract for Federal Government customers only.
For the most stringent laboratory requirements
Manufacturer: Eppendorf™ 022491148
 View more versions of this product
Catalog No. 05-403-06
  Check Availability
Add to Cart
    Specifications
    Product Type Pipet Tips
    Material Polypropylene
    Packaging Individually Wrapped, 100 pieces
    Length (English) 2.08 in.
    Length (Metric) 5.3cm
    Sterility Sterile
    Autoclavable Yes
    Tip Style Drop Contour Design, Extended, Finely Tapered, Universal Fit
    Certifications/Compliance ISO 9001
    Volume (Metric) 2 to 200μL
    Color Yellow
  • Description & Specifications
Related Products
Product Content Feedback