Eppendorf™ Combitips™ Plus Pipet Tips
Positive displacement for correct volumes
Manufacturer: Eppendorf™ 022266403
 View more versions of this product
Catalog No. 21-381-330
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type Specialty Tip
    Material Polypropylene
    For Use With (Equipment) For Eppendorf Repeater™ Plus pipetters
    Sterility Nonsterile
    Tip Style Positive Displacement
    Quantity Each
    Volume (Metric) 5mL
  • Description & Specifications
Related Products
Product Content Feedback