Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Combitips™ Plus Pipet Tips
Positive displacement for correct volumes
Specifications
Volume (Metric) | 0.5 mL |
---|---|
Tip Style | Specialty, Positive Displacement |
For Use With (Equipment) | Eppendorf Repeater, EDOS (Automatic Dispensing System) |
Product Line | CombiTips Plus |
Material | Polypropylene |
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
21-381-327
|
Eppendorf™
022266101 |
Nonsterile; Volume: 0.5mL; Dispensing range: 5 to 100μL | Nonsterile; Volume: 0.5mL; Dispensing range: 5 to 100μL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
21-381-336
|
Eppendorf™
022267205 |
Sterile; Volume: 0.5mL; Dispensing range: 5 to 100μL | Sterile; Volume: 0.5mL; Dispensing range: 5 to 100μL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
Description
Tips function according to the positive-displacement principle, ensuring that the correct volume is dispensed with regard to the density and viscosity of the liquid used. The positive displacement system provides safer dispensing of radioactive and toxic substances as well as solutions with high vapor pressure. No aerosols are formed.
- For use with Eppendorf Repeaters™ (original, Plus, Pro, stream and Xstream); and EDOS (automatic dispensing system)
- Available in nine volumes from 0.1 to 50mL
- Reliable results ensured by positive-displacement system
- Fine tip of Combitip provides exact dispensing without additional pipet tip
- Polyethylene plunger and polypropylene cylinder make these tips ideal for use with aggressive reagents
- Choose from sterile and nonsterile Combitips Plus Tips and Eppendorf Biopur™ tips, which are certified sterile as well as pyrogen-, RNase-, DNA-, and ATP-free
Specifications
0.5 mL | |
Eppendorf Repeater, EDOS (Automatic Dispensing System) | |
Polypropylene |
Specialty, Positive Displacement | |
CombiTips Plus |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title