Eppendorf™ Combitips™ Tips Available on GSA/VA Contract for Federal Government customers only.
For Eppendorf Repeater™ Pipetter
Manufacturer: Eppendorf™ 022261401
 View more versions of this product
Catalog No. 21-380-8C
  Check Availability
Add to Cart
    Specifications
    Product Type Specialty Tip
    Material Polypropylene cylinder; Polyethylene plunger
    For Use With (Equipment) For Eppendorf Repeater™ pipetter
    Sterility Nonsterile
    Quantity 100 Pack
    Volume (Metric) 12.5mL
  • Description & Specifications
Related Products
Product Content Feedback