Learn More
Eppendorf™ Combitips™ Tips Available on GSA/VA Contract for Federal Government customers only.
For Eppendorf Repeater™ Pipetter
Manufacturer:  Eppendorf™ 022260952
 View more versions of this product
Catalog No. 21-380-8J
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Specialty Tip
Nonsterile
50 Pack
0.05mL
Polypropylene cylinder; Polyethylene plunger
For Eppendorf Repeater™ pipetter