Learn More
Eppendorf™ Combitips™ Tips Available on GSA/VA Contract for Federal Government customers only.
For Eppendorf Repeater™ Pipetter
Manufacturer:  Eppendorf™ 022495101
 View more versions of this product
Catalog No. 21-381-307B
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Specialty Tip, Biopur
Sterile
100 Case
1.25mL
Polypropylene cylinder; Polyethylene plunger
For Eppendorf Repeater™ pipetter