Learn More
Eppendorf™ DNA LoBind Microcentrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Perfect for DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology
Manufacturer:  Eppendorf™ 925000064
 View more versions of this product
Catalog No. 13-864-254
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
DNA, DNase, RNase, PCR Inhibitors free
1.5mL
Polypropylene
No
1000
Safe-lock lid
Clear
30000xG
DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology
Conical