Eppendorf™ DNA LoBind Microcentrifuge Tubes
Perfect for DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology
Manufacturer: Eppendorf™ 925000066
 View more versions of this product
Catalog No. 13-864-255
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type DNA Lobind Microcentrifuge Tube
    Capacity (Metric) 2mL
    Color Clear
    Material Polypropylene
    Max. Force 25000xG
    Sterile No
    For Use With (Application) DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology
    Packaging 1000
    Certifications/Compliance DNA, DNase, RNase, PCR Inhibitors free
    Shape Conical
    Closure Type Safe-lock lid
  • Description & Specifications
Related Products
Product Content Feedback