Learn More
Eppendorf™ epT.I.P.S.™ Pipette Tips, Racks, Eppendorf Biopur™, 50-1250μL
Single-use pipette tips for aspirating and dispensing liquids with air cushion pipettes. Eppendorf™ epT.I.P.S.™ Pipette Tips feature packaging that is color coded and corresponds to the color of the control button of the respective Eppendorf pipette.
Manufacturer:  Eppendorf™ 022494014
Catalog No. 05412305
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Eppendorf Biopur offers the highest biological purity grade
  • Guaranteed PCR clean, sterile, pyrogen, ATP, and DNA free
  • For use in medical, pharmaceutical, food industry, molecular biology, and cell technology
  • Each production lot inspected continuously by independent laboratory
Specifications
Specifications
Dark green
Autoclavable
10.3cm
50 to 1250 μL
PP (Polypropylene)
Rack