Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ SealMax Filter Pipet Tips
Tips with liquid blocking technology provide additional protection against over pipetting
Supplier: Eppendorf™ 0030077822
This item is not returnable.
View return policy
Description
- Sterile tips made of polyethylene
- White and violet filter layers in the tip retain aerosols and biomolecules
- Purple filter layer blocks liquids, binds aerosols and does not allow any liquid to pass through
- Hydrophobic white layer repels drops, splashes and binds aerosols
- Free of PCR inhibiting additives
- Provide protection against contamination of pipet and sample
- Color coded tip boxes simplify matching pipetes with their tips
PCR and microbiology procedures
Order Info
10 Racks x 96 tips
Specifications
2 to 100 μL | |
Sterile | |
5.3cm | |
ep DualFilter T.I.P.S |
Filtered | |
Yellow | |
Rack | |
Polyethylene |