Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ SealMax Filter Pipet Tips Available on GSA/VA Contract for Federal Government customers only.
Tips with liquid blocking technology provide additional protection against over pipetting
Manufacturer:  Eppendorf™ 0030077806
 View more versions of this product
Catalog No. 13-683-750
  Check Availability
Add to cart
Specifications
Description
  • Sterile tips made of polyethylene
  • White and violet filter layers in the tip retain aerosols and biomolecules
  • Purple filter layer blocks liquids, binds aerosols and does not allow any liquid to pass through
  • Hydrophobic white layer repels drops, splashes and binds aerosols
  • Free of PCR inhibiting additives
  • Provide protection against contamination of pipet and sample
  • Color coded tip boxes simplify matching pipetes with their tips
PCR and microbiology procedures
Description & Specifications
Specifications
Filter Pipet Tips
Sterile
Polyethylene
3.4cm
0.1 to 10μL
Dark Gray
960 Pack