Eppendorf™ epTIPS™ Filter Tips Available on GSA/VA Contract for Federal Government customers only.
Ideal for small volume pipetting
Manufacturer: Eppendorf™ 022491296
 View more versions of this product
Catalog No. 04-987-420
  Check Availability
Add to Cart
    Specifications
    Product Type Drop Contour Tip
    Material Polyethylene
    Length (English) 2.16 in.
    Length (Metric) 5.5cm
    Sterility Sterile
    Volume (Metric) 2 to 200μL
    Packaging Rack
    Certifications/Compliance ISO 9001
    Color Yellow
  • Description & Specifications
Related Products
Product Content Feedback