Learn More
Eppendorf™ epTIPS LoRetention Dualfilter Tips, PCR Clean Available on GSA/VA Contract for Federal Government customers only.
Increase reproducibility and and notably reduce the loss of expensive reagents
Manufacturer:  Eppendorf™ 022493000
 View more versions of this product
Catalog No. 02-717-340
  Check Availability
Add to cart
Description
Description
The surface of epT.I.P.S. LoRetention is achieved through an innovative treatment at the molecular level: the Pearl Effect Technology. The tips are uncoated and free from additives, thus nothing can bleed into the sample. Liquids roll off completely, so that almost no sample remains in the tip. In comparison, the adhesive layer of liquid in standard tips retains considerably more sample material. This unique material property guarantees maximum sample recovery, improved reproducibility and higher sensitivity.
  • Ultra hydrophobic and homogeneous surface for maximum reproducibility
  • Ultra-hydrophobic surface for minimum loss of sample
  • Air bubbles and foaming are significantly reduced during pipetting
  • Autoclavable with high chemical resistance
  • Shelf life is 5 years
  • Available as ep Dualfilter T.I.P.S. and in PCR clean purity grade
  • Universal tip fit
PCR and real-time PCR preparation of enzyme solutions and master mixes, Isolation, purification and denaturation of proteins, Restriction digestions and ligations, Pipetting DNA ladders during gel electrophoresis
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Rack
0.1 to 10μL
Sterile
Yes
Low Retention
Filtered
Dark Gray
PCR and real-time PCR preparation of enzyme solutions and master mixes, isolation, purification and denaturation of proteins, restriction digestions and ligations, pipetting DNA ladders during gel electrophoresis