Eppendorf™ epTIPS LoRetention Dualfilter Tips, PCR Clean Available on GSA/VA Contract for Federal Government customers only.
Increase reproducibility and and notably reduce the loss of expensive reagents
Manufacturer: Eppendorf™ 022493004
 View more versions of this product
Catalog No. 02-717-342
  Check Availability
Add to Cart
    Specifications
    Product Type LoRetention Dualfilter Tip
    Packaging 10 x 96/Rack
    Sterility Sterile
    Autoclavable Yes
    For Use With (Application) PCR and real-time PCR preparation of enzyme solutions and master mixes, isolation, purification and denaturation of proteins, restriction digestions and ligations, pipetting DNA ladders during gel electrophoresis
    Volume (Metric) 20 to 300μL
    Color Yellow
  • Description & Specifications
Related Products
Product Content Feedback