Eppendorf™ epTIPS™ LoRetention Tips Available on GSA/VA Contract for Federal Government customers only.
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
Manufacturer: Eppendorf™ 0030072286
 View more versions of this product
Catalog No. 13-683-504
  Check Availability
Add to Cart
    Specifications
    Product Type LoRetention Tip
    Length (English) 2.79 in.
    Length (Metric) 7.1cm
    Autoclavable Yes
    For Use With (Application) Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
    Volume (Metric) 50 to 1000μL
    Color Blue
  • Description & Specifications
Related Products
Product Content Feedback