Eppendorf™ epTIPS LoRetention Tips, PCR Clean Reloads

Increase reproducibility and and notably reduce the loss of expensive reagents

Manufacturer: Eppendorf™ 022493028

 View all options for this product

Catalog No. 02-717-354


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Product Type LoRetention Tip
    Volume (Metric) 0.5 to 20μL
    Sterility Sterile
    Color Light Gray
    Autoclavable Yes
    Packaging PCR Clean Racks
    For Use With (Application) Preparation of enzyme solutions and master mixes, isolation, purification and denaturation of proteins, restriction digestions and ligations, pipetting DNA ladders during gel electrophoresis

    The surface of epT.I.P.S. LoRetention is achieved through an innovative treatment at the molecular level: the Pearl Effect Technology. The tips are uncoated and free from additives, thus nothing can bleed into the sample. Liquids roll off completely, so that almost no sample remains in the tip. In comparison, the adhesive layer of liquid in standard tips retains considerably more sample material. This unique material property guarantees maximum sample recovery, improved reproducibility and higher sensitivity.

    • Ultra hydrophobic and homogeneous surface for maximum reproducibility
    • Ultra-hydrophobic surface for minimum loss of sample
    • Air bubbles and foaming are significantly reduced during pipetting
    • Autoclavable with high chemical resistance
    • Shelf life is 5 years
    • Universal tip fit

    PCR and real-time PCR preparation of enzyme solutions and master mixes, Isolation, purification and denaturation of proteins, Restriction digestions and ligations, Pipetting DNA ladders during gel electrophoresis

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit