Eppendorf™ epTIPS™ PCR Clean Available on GSA/VA Contract for Federal Government customers only.
For all molecular biology applications
Manufacturer: Eppendorf™ 022491717
 View more versions of this product
Catalog No. 05-403-47
  Check Availability
Add to Cart
    Specifications
    Sterility Sterile
    Product Type PCR Clean Reload Tip
    Length (English) 1.57 in.
    Length (Metric) 4cm
    Autoclavable Yes
    Tip Style Drop Contour Design, Universal Fit
    Packaging Refill Trays
    Certifications/Compliance ISO 9001
    Volume (Metric) 0.1 to 20μL
    Color Gray
  • Description & Specifications
Related Products
Product Content Feedback