Eppendorf™ epTIPS™ PCR Clean Available on GSA/VA Contract for Federal Government customers only.
For all molecular biology applications
Manufacturer: Eppendorf™ 022491733
 View more versions of this product
Catalog No. 05-403-49
  Check Availability
Add to Cart
    Specifications
    Product Type PCR Clean Reload Tip
    Length (English) 2.08 in.
    Length (Metric) 5.3cm
    Sterility Sterile
    Autoclavable Yes
    Tip Style Drop Contour Design, Universal Fit
    Packaging Refill Trays
    Certifications/Compliance ISO 9001
    Volume (Metric) 2 to 200μL
    Color Yellow
  • Description & Specifications
Related Products
Product Content Feedback