Eppendorf™ epTIPS, Reloads

System optimized for use with multichannel pipettes

Manufacturer: Eppendorf™ 022491540

 View all options for this product

Catalog No. 13-684-277


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Product Type Reload Tip
    Volume (Metric) 2 to 100μL
    Color Yellow
    Autoclavable Yes
    Length (English) 2.08 in.
    Length (Metric) 5.3cm
    Certifications/Compliance ISO 9001:2000

    Tips can be attached to the pipette from the refill trays. Refill trays are entirely autoclavable for future use.

    • Drop contour design
    • Finely tapered and extended; ideal for small-volume pipetting Fine graduations make it easy to check volume Soft, flexible tip cone provides better seal to pipetter

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit