Eppendorf™ epTIPS, Reloads
System optimized for use with multichannel pipettes
Manufacturer: Eppendorf™ 022491505
 View all options for this product
Catalog No. 13-684-271
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Volume (Metric) 0.1-10μL
    Tips can be attached to the pipette from the refill trays. Refill trays are entirely autoclavable for future use.
    • Drop contour design
    • Finely tapered and extended; ideal for small-volume pipetting Fine graduations make it easy to check volume Soft, flexible tip cone provides better seal to pipetter
  • Description & Specifications
Related Products
Product Content Feedback