Eppendorf™ FA-45-20-17 Rotor Adapters
For 1.5/2mL tubes, cryotubes and 12mm HPLC vials
Manufacturer: Eppendorf™ 5820769009
 View more versions of this product
Catalog No. 58-207-69009
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Tube Type Cryotubes
    For Use With (Equipment) FA-45-20-17 (1 x 5mL) rotor in Model 5804/5810 centrifuges
  • Description & Specifications
Related Products
Product Content Feedback