Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ FA-45-6-30 Rotor Adapters
For volumes up to 50mL
Supplier: Eppendorf™ 5820730005
Description
- FA-45-6-30 Rotor Adapters
- For volumes up to 50mL
Specifications
Rotor Adapter | |
1 x 5 mL Conical Tube | |
Rotor for 5804/5810 centrifuge | |
1 Tube |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction