Eppendorf™ IsoTherm Systems: Replacement Components for Starter Sets Available on GSA/VA Contract for Federal Government customers only.

Protect temperature-sensitive biochemical and microbiological samples while working at the lab bench

Manufacturer: Eppendorf™ 022510207

 View more versions of this product

Catalog No. 05-405-17

  Check Availability
Add to Cart

    Specifications

    Product Type IsoPack Set
    For Use With (Equipment) For use with the Eppendorf IsoTherm system
    Holds 1.5 and 2mL tube at -21°C for up to 3 Hours
    For Use With (Application) Used for cooling samples effectively and consistently at -21°C or 0°C over many Hours
    Material Polycarbonate
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit