Learn More
Eppendorf™ Microcapillaries for ICSI
Disposable glass capillaries for handling sperm and stem cells
Specifications
Material Glass
Sterility Sterile
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Type Includes Price Quantity & Availability  
E5175107004
View Documents
Eppendorf™
930001040
15 μm 20 μm Transfer Tip Rigid 1mm flange This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
TransferTip-F
  • Designed for injecting human sperm
  • Heat-formed spike facilitates penetration of oocytes
  • Feature a 4µm inner diameter, 7µm outer diameter, flexible 0.4mm flange and 35° capillary angle
TransferTip-ES
  • Designed for transferring stem cells into blastocysts
  • Heat-formed spike facilitates penetration of blastocysts
  • Features a 20mm inner diameter, rigid 1m flange and 2° capillary angle
VacuTip
  • Designed to hold suspension cells (e.g., oocytes or blastocysts)
  • Features 15µm inner diameter, 100µm outer diameter, 1mm flange, and 35° capillary angle
Specifications
Specifications
Glass
Sterile