Eppendorf™ Model 5424 Microcentrifuge Rotors Available on GSA/VA Contract for Federal Government customers only.
For use with Eppendorf Model 5424 Microcentrifuges
Manufacturer: Eppendorf™ 022653008
 View more versions of this product
Catalog No. 05-400-008
  Check Availability
Add to Cart
    Specifications
    Type Standard rotor
    Includes Aerosol-tight lid
    No. of Positions 24.0
    Capacity (Metric) 24 x 1.5/2mL
    Hertz 50/60Hz
    Speed 15,000rpm
    Radius (English) 3.3 in.
    Radius (Metric) 8.4cm
  • Description & Specifications
Related Products
Product Content Feedback