Eppendorf™ Model 5430 Microcentrifuge Rotor Accessory, Adapters Available on GSA/VA Contract for Federal Government customers only.
For use with Eppendorf Model 5430 Microcentrifuges and compatible rotors
Manufacturer: Eppendorf™ 022654349
 View more versions of this product
Catalog No. 05-401-515
  Check Availability
Add to Cart
    Specifications
    Type Adapter
    Product Type Adapter
    Capacity (Metric) 29.5 x 100-125mm/50mL Falcon tubes (set of 2)
    Tube Size 29.5mm
    Style 29.5 x 100-125mm/50mL Falcon™ tubes
    For Use With (Equipment) Rotor
    Angle Fixed
  • Description & Specifications
Related Products
Product Content Feedback