Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ PCR-Strip Rotor

For use with 5417 Series centrifuges

Manufacturer:  Eppendorf™ 022636146

Catalog No. 05-40-642

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ideal rotor for centrifuging PCR tubes.

  • Accepts 6 × 8-tube strips or 48 × 0.2mL PCR tubes
  • Maximum speed/RCF: 10,000rpm/10,600 x g

Specifications

Specifications

Rotor
10,600 x g
5430 Microcentrifuge
48 x 0.2 mL
10,000 rpm
6 x 8 Tube strip, 6 x 5 tube strip
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit