Eppendorf™ Refrigerated Multipurpose Centrifuge 5804 R Rotor Adapters Available on GSA/VA Contract for Federal Government customers only.
For 6 x 85mL fixed-angle rotors
Manufacturer: Eppendorf™ 022637223
 View more versions of this product
Catalog No. 05-400-83
  Check Availability
Add to Cart
    Specifications
    Device Holds 2
    Tube Size 4 - 10mL
    Bore Size (Metric) 1.6cm
    For Use With (Equipment) Eppendorf™ special rotor for 5804/5804R/5810/5810R microcentrifuge
  • Description & Specifications
Related Products
Product Content Feedback