Learn More
Eppendorf™ Repeater™ M4 Pipetes
Unique volume range of 1µLto 10mL
Manufacturer:  Eppendorf™ A14H149820
Catalog No. 05-412-563
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Uses positive displacement principal to precisely and accurately dispense virtually any liquid
  • Dispenses aspirated liquid up to 100 times without refilling the tip
  • Step counter supports error-free working
  • Has 20 different volume settings for every Combitip size
  • Automatic Combitip recognition and volume display
  • Ergonomically shaped operating lever to dispense liquid and eject Combitip advanced
  • Ergonomically shaped filling lever for filling and emptying Combitip advanced
  • Hand rest supports relaxed hand position
  • Repeater switches off when not in use, and back on with minimal movements to save power and time
Applications:
  • Repetitive Dispensing of volumes in long series such as tube or plate filling, aliquoting reagents and kit usage
  • Contamination-free dispensing of toxic, radioactive and or other hazardous liquids
  • Precise and accurate dispensing of viscous solutions